Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Hektoen Enteric Agar Plate

Catalog No. EBP3190
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P3190 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P3190 Supplier Edge Biologicals Supplier No. P3190
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Proteose Peptone 12g/L, Yeast extract 3g/L, Bile Salts #3 9g/L, Lactose 12g/L, Saccharose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Bromthymol blue 0.065g/L, Acid Fuchsin 0.1g/L, Agar 14g/L, Deionized

Effective testing agent for Salmonella and Shigella

Used in the examination of clinical materials and food specimens.

  • Prepared agar plate suspension
  • Color: Green to dark green
  • Sterile
  • Autoclavable
  • Final pH is 7.53

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Green
Content And Storage 8°C to 12°C
Detects Salmonella, Shigella
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Hektoen enteric Agar
Sterility Sterile
Formulation Peptone - 12 g, NaCl - 5 g, Yeast Extract - 3 g, Sodium Thiosulfate - 5 g, Bile Salts No. 3 - 9 g, Ferric Ammonium Citrate - 1.5 g, Lactose - 12 g, Bromthymol Blue - 0.065 g, Sucrose - 12 g, Acid Fuchsin - 0.1 g, Salicin - 2 g, Agar - 14.0 g, Distilled Water - 1 L
For Use With (Application) Isolation and cultivation of gram negative microorganisms, particularly Salmonella and Shigella
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.