Learn More
Edge Biologicals Streptococcus faecalis Broth
Catalog No. EBT0666
Change view
Click to view available options
Quantity:
5 mL, 10/Pk.
Catalog No. Quantity
EB-T0666 5 mL, 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0666 Supplier Edge Biologicals Supplier No. T0666
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective medium
Streptococcus faecalis Broth (SF) is a selective medium that is used for the isolation of fecal Streptococci.
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Media Type Streptococcus faecalis Broth (SF)
Quantity 5 mL, 10/Pk.