Learn More
Edge Biologicals Litmus Milk Medium
Catalog No. EBT0311
Change view
Click to view available options
9mL; 10/Pk.
Catalog No.
EB-T0311 9mL; 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0311 Supplier Edge Biologicals Supplier No. T0311
This item is not returnable. View return policy
9mL
Litmus Milk Medium is used for the maintenance of Lactic Acid Bacteria as well as to determine the identity of bacteria based on their ability to metabolize milk.
Specifications
Media Type Litmus Milk Medium
Quantity 10/Pk.