Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals MacConkey Agar Plate

Catalog No. EBP1205
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P1205 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P1205 Supplier Edge Biologicals Supplier No. P1205
Add to Cart
Add to Cart

Selective and differential plating medium

  • Used in the isolation and cultivation of gram-negative organisms, especially coliforms and enteric pathogens from clinical and non-clinical specimens
  • Color: purplish red
  • Sterile, autoclavable
  • pH after sterilization: 7.25

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Purple
Content And Storage 8°C to 12°C
Form Liquid
Manufacturing Origin United States
Media Type MacConkey Agar Plate
Sterility Sterile
Formulation Peptone 17g/L, Proteose Peptone 3g/L, Lactose 10g/L, Bile Salts #3 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.05g/L, Crystal Violet 0.001g/L, Agar 13.5g/L, Deionized Water 1000mL
For Use With (Application) Isolation and cultivation of gram negative species, particularly Coliforms and enteric pathogens from clinical or nonclinical specimens
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.