Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals MacConkey Sorbitol Agar Plate

Catalog No. EBP4376
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-P4376 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-P4376 Supplier Edge Biologicals Supplier No. P4376
Add to Cart
Add to Cart

Selective medium

MacConkey Sorbitol Agar is used for the isolation and cultivation of enteropathogenic Escherichia coli serotypes from clinical specimens and foods.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Detects Enteropathogenic Escherichia coli serotypes
Form Prepared Medium
Format Agar Plate
Media Type MacConkey Sorbitol Agar
Product Type Agar Plate
Target Organism Class Bacteria
Test Type Microbial Cultivation
For Use With (Application) Isolation and cultivation of enteropathogenic Escherichia coli serotypes
Packaging Type Pack
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.