Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Mannitol Salt Agar Plate

Catalog No. EBP2210
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P2210 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P2210 Supplier Edge Biologicals Supplier No. P2210
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the selective isolation and enumeration of staphylococci from clinical and nonclinical materials

  • Culture for isolating staphylococci
  • Sterile
  • Autoclavable
  • Format: Suspension
  • Color: Clear reddish pink
  • pH after Sterilization: 7.31

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Clear reddish pink
Content And Storage 8°C to 12°C
Form Agar Suspension
Manufacturing Origin United States
Media Type Mannitol Salt Agar
Sterility Sterile
Formulation Mannitol Salt, Peptone #3 10g/L, Beef extract 1g/L, Mannitol 10g/L, Sodium Chloride 75g/L, Agar 16g/L, Phenol red 25mg/L, Crystal Violet 2mg/L, Deionized Water 1000mL
For Use With (Application) Isolating Staphylococci
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.