Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Middlebrook 7H11 Agar Slant

Catalog No. EBT0366
Change view
Click to view available options
Middlebrook 7h9 Broth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0366 Middlebrook 7h9 Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0366 Supplier Edge Biologicals Supplier No. T0366
This item is not returnable. View return policy

For the isolation of mycobacteria, specifically Mycobacterium tuberculosis

  • Sterile
  • Autoclavable

Specifications

Format Slant
Manufacturing Origin United States
Media Type Middlebrook 7H11 Agar
For Use With (Application) Detection of mycobacteria
Packaging Type Tube
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.