Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Motility Test Medium Tube

Catalog No. EBT0391
Change view
Click to view available options
Motility Test Medium Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0391 Motility Test Medium Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0391 Supplier Edge Biologicals Supplier No. T0391

Used in the determination of bacterial motility

  • Color: clear tan
  • Sterile, autoclavable
  • Final pH: 7.39

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance M22-A3
Color Tan
Manufacturing Origin United States
Product Type Motility Test Medium
For Use With (Application) Determination of bacterial motility and bacterial differentiation
Packaging Type Tube
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.