Learn More
Edge Biologicals Mueller Hinton Agar Plate
Catalog No. EBLP100
Change view
Click to view available options
10/Pk.
Catalog No.
EB-LP100 10/Pk.
1 options
Catalog No. EB-LP100 Supplier Edge Biologicals Supplier No. LP100
Used for testing the susceptibility of microorganisms to antimicrobial agents
This medium is used in the disc diffusion technique in CLSI Approved Standard:M100-S17, Performance Standards for Antimicrobic Disc Susceptibility Tests.
Specifications
Color light amber
Format Plate
Media Type Mueller Hinton Agar
Includes Beef Extract 2g, Acid Hydrolysate of Casein 17.5g, Starch 1.5g, Agar 17g, Deionized water 1000mL
Quantity 10/Pk.