Learn More
Edge Biologicals Nutrient Agar Plate
Description
Clear amber agar plate has a pH of 6.84 ( 6.8 ±0.2) after sterilization. For the cultivation of the majority of less fastidious microorganisms.
Composition (g/L):
- Beef Extract, 3g
- Peptone, 5g
- Sodium chloride, 8g
- Agar, 15g
- Deionized water, 1000mL
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis |
| Color | Light Amber |
| Content And Storage | 8°C to 12°C |
| Detects | Fastidious Microorganisms |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Nutrient Agar |
| Sterility | Sterile |
| Formulation | Beef extract 3g/L, Peptone 5g/L, Sodium Chloride 8g/L, Agar 15g/L, Deionized Water 1000mL |
| For Use With (Application) | Cultivation of many of the less fastidious microorganisms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.