Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Phenylethyl Alcohol (PEA) Agar Plate

Catalog No. EBP3275
Change view
Click to view available options
Phenylethyl Alcohol (PEA) Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-P3275 Phenylethyl Alcohol (PEA) Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P3275 Supplier Edge Biologicals Supplier No. 3P275
This item is not returnable. View return policy

Selective medium used in the growth of gram-positive cocci from clinical specimens

  • Color: Light amber
  • Sterile
  • Autoclavable
  • Format: Suspension
  • Final pH is 7.25
  • Does not include sheep's blood

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Color Light Amber
Manufacturing Origin United States
Media Type Phenylethyl Alcohol Agar
Sterility Sterile
For Use With (Application) Cultivation of gram positive bacteria
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.