Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Plated Culture Media: Blood Agar/Chocolate Agar

Catalog No. EBP3014
Change view
Click to view available options
Blood Agar/Chocolate Agar; 15 x 100mm; Container: Biplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-P3014 Blood Agar/Chocolate Agar; 15 x 100mm; Container: Biplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P3014 Supplier Edge Biologicals Supplier No. P3014
This item is not returnable. View return policy

For detection of Neisseria/Hemophilus

Specifications

Form Dehydrated
Media Type Blood Agar/Chocolate Agar
For Use With (Application) Detection of Neisseria/Hemophilus
Packaging Type Biplate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.