Learn More
Edge Biologicals Sabouraud Dextrose Agar Plate
Catalog No. EBP4320
Change view
Click to view available options
Color:
light amber
Catalog No. Color
EB-P4320 light amber
1 options
This item is not returnable. View return policy
Catalog No. EB-P4320 Supplier Edge Biologicals Supplier No. P4320
Add to Cart
Add to Cart
This item is not returnable. View return policy
Recommended for the isolation of pathogenic yeasts and fungi from clinical specimens
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Color light amber
Format Plate
Media Type Sabouraud Dextrose Agar
Includes Bacto Neopepetone 10g, Dextrose 20g, Agar 15g, Deionized water 1000mL
Quantity 10/Pk.