Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Salmonella Shigella (SS) Agar Plate

Catalog No. EBP2380
Change view
Click to view available options
Certifications/Compliance:
Certificate of analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
EB-P2380 Certificate of analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-P2380 Supplier Edge Biologicals Supplier No. P2380
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in the isolation of Salmonella and Shigella spp. from clinical specimens

  • Selective and differential medium
  • Color: Light rose tan
  • Format: Solid
  • Sterile
  • Autoclavable
  • Final pH is 6.99

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance Certificate of analysis
Color Light Rose Tan
Content And Storage 8-25°C
Form Liquid
Manufacturing Origin United States
Media Type Salmonella Shigella Agar
Sterility Sterile
Formulation Beef extract 5g/L, Enzymatic digest of Casein 2.5g/L, Sodium Thiosulfate 8.5g/L, Enzymatic digest Animal tissue 2.5g/L, Bile Salts 8.5g/L, Lactose 10g/L, Sodium Citrate 8.5g/L, Ferric Citrate 1g/L, Brilliant green 0.33mg/L, Neutral red 25mg/L, Agar 13.5g/L, Deionized Water 1000mL
For Use With (Application) Isolation of Salmonella and Shigella from clinical and nonclinical samples
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.