Learn More
Edge Biologicals Selective Buffered Charcoal Yeast Extract Agar
Catalog No. EBP4129
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
EB-P4129 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. EB-P4129 Supplier Edge Biologicals Supplier No. P4129
Add to Cart
Add to Cart
This item is not returnable. View return policy
For primary isolation and cultivation of Legionella pneumophila and other Legionella species from clinical and environmental samples
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Quantity 10/Pk.