Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Thioglycollate Broth

Catalog No. EBT0714
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EB-T0714 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EB-T0714 Supplier Edge Biologicals Supplier No. T0714
Add to Cart
Add to Cart

For cultivation and detection of microorganisms, including bacteria and anaerobes

  • Prepared liquid broth
  • Colors: Clear, pale amber
  • Sterile, autoclavable
  • Volume: 10mL
  • Final pH is 7.15

For cultivation and detection of microorganisms, including bacteria and anaerobes

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Certifications/Compliance M22-A3
Color Light Amber
Content And Storage 8° to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Broth
Formulation Pancreatic digest of Casein 17g/L, digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, L-Cystine 0.25g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, Sodium Sulfite 0.1g/L, Deionized Water 960mL
For Use With (Application) Cultivation of obligate anaerobes, facultative anaerobes, microaerophiles or for detection of bacteria in normally sterile specimens
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.