Learn More
Edge Biologicals Triple Sugar Iron Slant
Description
- Selective medium
- Color: deep orange to red orange
- Sterile, autoclavable
- Final pH is 7.40
- 10/Pk.
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
| Certifications/Compliance | M22-A3 |
| Color | Deep Orange |
| Content And Storage | 8°C to 12°C |
| Detects | Enterobacteriaceae |
| Form | Suspension |
| Format | Tube |
| Manufacturing Origin | United States |
| Media Type | Triple Sugar Iron Agar |
| Formulation | Beef extract 3g/L, Pancreatic digest Casein 15g/mL, Dextrose 1g/L, Sucrose 10g/mL, Sodium Chloride 5g/mL, Agar 12g/mL, De-ionized Water 1000mL, Yeast extract 3g/mL, Proteose Peptone #3 5g/mL, Lactose 10g/mL, Ferrous Sulfate 0.2g/mL, Sodium Thiosulfate 0.3g/mL, Phenol red 24mg/L |
| For Use With (Application) | For identification of Enterobacteria based on fermentation of Glucose, Lactose and Sucrose as well as Hydrogen Sulfide production |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.