Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Tryptic Soy Broth with 6.5% NaCl

Catalog No. EBT0762
Change view
Click to view available options
Manufacturing Origin:
United States
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Manufacturing Origin
EB-T0762 United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0762 Supplier Edge Biologicals Supplier No. T0762
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile and autoclavable liquid used to differentiate the enterococcal group D from non-enterococcal group D streptococci

Amber-colored broth has a final pH of 7.15.

Composition (g/L):

  • Pancreatic digest of casein, 17g
  • Enzymatic digest of soybean meal, 3g
  • Sodium chloride, 5g
  • Dextrose, 2.5g
  • Dipotassium phosphate, 2.5g
  • Deionized water, 1000mL

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Manufacturing Origin United States
Media Type Tryptic Soy Broth
For Use With (Application) To differentiate the enterococcal group D from non-enterococcal group D streptococci by 6.5% NaCl tolerance test
Quantity 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.