Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Tryptic Soy Broth (TSB) with 15% glycerol

Catalog No. EBT0764
Change view
Click to view available options
2mL, 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0764 2mL, 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0764 Supplier Edge Biologicals Supplier No. T0764
This item is not returnable. View return policy

Maintenance medium

Tryptic Soy Broth (TSB) with glycerol (15%) is used in the long-term frozen maintenance of bacterial stock cultures.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Media Type Tryptic Soy Broth (TSB) with glycerol (15%)
Quantity 2 mL, 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.