Learn More
Edge Biologicals TSA W/ 5% Sheep Blood Plate
Catalog No. EBP2075
Change view
Click to view available options
10/Pk.
Catalog No.
EB-P2075 10/Pk.
1 options
Catalog No. EB-P2075 Supplier Edge Biologicals Supplier No. 2P075
General purpose medium
Tryptic soy agar w/ 5% sheep blood (Blood agar) is recommended for use as a general purpose medium for the growth of a wide variety of microorganisms and for determining hemolytic reactions.
Specifications
Media Type TSA W/ 5% Sheep Blood
Quantity 10/Pk.