Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Urea Agar Slant

Catalog No. EBT0771
Change view
Click to view available options
Urea Agar Slant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0771 Urea Agar Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0771 Supplier Edge Biologicals Supplier No. T0771
This item is not returnable. View return policy

Detection medium for urease activity of bacteria, esp. Proteus and Providencia species

  • Sterile, autoclavable
  • Final pH is 6.90
  • Color: clear tan

For the detection of urease activity of bacteria, especially members of Proteus and Providencia species.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Content And Storage 8° to 25°C
Form Slant
Manufacturing Origin United States
Formulation Peptone 1g/L, Glucose 1g/L, Sodium Chloride 5g/L, Disodium phosphate 1.2g/L, Potassium dihydrogen phosphate 0.8g/L, Phenol red 0.012g/L, Agar 15g/L
For Use With (Application) For preparation of Christensen Agar medium, detection of Urease activity of bacteria including Proteus and Providencia species
Packaging Type Tube
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.