Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edge Biologicals Water Blanks (Butterfields Blanks)

Catalog No. EBT0782
Change view
Click to view available options
9mL Plastic; Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EB-T0782 9mL Plastic; Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. EB-T0782 Supplier Edge Biologicals Supplier No. T0782
Add to Cart
Add to Cart
This item is not returnable. View return policy

Diluent for detecting pathogenic microorganisms in food and environmental sources.

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Cap Type Screw Cap
Certifications/Compliance AOAC Recommended
Detects Pathogenic Microorganisms
Format Pre-filled Tube
Media Type Butterfields Blanks
Test Type Microbial Detection
Fill Volume 9 mL
Formulation Butterfield's Phosphate Buffer (BPB)
For Use With (Application) Detecting Pathogenic Microorganisms, Microbiological Determinations
Includes 10 Plastic Screw Cap Tubes, Each with 9 mL Butterfield's Phosphate Buffer
Material Plastic
Packaging Type Pack
Quantity 9 mL Plastic, 10/Pk.
Specimen Type Raw Specimens, Food, Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.