Learn More
Edge Biologicals Citrate Agar Slant
Catalog No. EBT0130
Change view
Click to view available options
10/Pk.
Catalog No.
EB-T0130 10/Pk.
1 options
Catalog No. EB-T0130 Supplier Edge Biologicals Supplier No. T0130
Differential medium
Simmon's Citrate Agar is used for the differentiation of Enterobacteriaceae, based on the utilization of sodium citrate.
Specifications
Media Type Simmon's Citrate Agar
Quantity 10/Pk.