Learn More
MSC EDGE CR GLOVE
Catalog No. 5000687614
Encompass
Ansell Cut, Puncture & Abrasion-Resistant Gloves are designed to provide hand protection in demanding work environments. Constructed from polyethylene and fiberglass with a padded leather palm grip, these gray, full-fingered knit gloves offer good dexterity and flexibility.
  • Medium cut protection and high levels of abrasion
  • Leather pad enhances grip, cut, and abrasion protection
  • Provides cushioning during stamping applications
  • Good grip in dry and slightly oily environments
  • Gauge: 13
  • ANSI/ISEA cut resistance level: A4
  • ANSI/ISEA puncture resistance level: 4
  • ANSI/ISEA abrasion resistance level: 4
Catalog No. 50-006-87614 Supplier MSC Supplier No. 41305228
Ansell Cut, Puncture & Abrasion-Resistant Gloves are designed to provide hand protection in demanding work environments. Constructed from polyethylene and fiberglass with a padded leather palm grip, these gray, full-fingered knit gloves offer good dexterity and flexibility.
  • Medium cut protection and high levels of abrasion
  • Leather pad enhances grip, cut, and abrasion protection
  • Provides cushioning during stamping applications
  • Good grip in dry and slightly oily environments
  • Gauge: 13
  • ANSI/ISEA cut resistance level: A4
  • ANSI/ISEA puncture resistance level: 4
  • ANSI/ISEA abrasion resistance level: 4