Learn More
MSC Edge Eyewear Reclus, Smoke Lens, ANSI Z87.1 Compliant, 1 ea
Catalog No. 5000675686
Encompass
The Reclus safety glasses are designed for job site use and demanding outdoor activities, offering a stylish, comfortable, and protective eyewear solution. They feature a wraparound design for improved peripheral vision and enhanced protection.
  • Complies with ANSI Z87.1+ standards
  • Frame constructed of flexible, durable nylon composite
  • Thermoplastic rubber nosepieces for enhanced comfort and anti-slip
  • Lenses block 99.9% of UVA/UVB/UVC rays
  • Straight temples with Tapered Lens Technology (TLT)
Catalog No. 50-006-75686 Supplier MSC Supplier No. 81945891
The Reclus safety glasses are designed for job site use and demanding outdoor activities, offering a stylish, comfortable, and protective eyewear solution. They feature a wraparound design for improved peripheral vision and enhanced protection.
  • Complies with ANSI Z87.1+ standards
  • Frame constructed of flexible, durable nylon composite
  • Thermoplastic rubber nosepieces for enhanced comfort and anti-slip
  • Lenses block 99.9% of UVA/UVB/UVC rays
  • Straight temples with Tapered Lens Technology (TLT)