Learn More
MSC Edge Eyewear Safety Glasses, Anti-Fog, Polycarbonate, Polarized, Smoke, 1 ea
Catalog No. 5000684279
Encompass
These safety glasses feature anti-fog, polycarbonate, smoke lenses in a full-framed, wrap-around design, providing essential eye protection with enhanced visibility.
  • Anti-fog coating
  • Polycarbonate smoke lenses
  • Full-framed, wrap-around design
  • Polarized for glare reduction
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Black nylon frame
  • Non-adjustable temples
  • Universal size
Catalog No. 50-006-84279 Supplier MSC Supplier No. 61497426
These safety glasses feature anti-fog, polycarbonate, smoke lenses in a full-framed, wrap-around design, providing essential eye protection with enhanced visibility.
  • Anti-fog coating
  • Polycarbonate smoke lenses
  • Full-framed, wrap-around design
  • Polarized for glare reduction
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Black nylon frame
  • Non-adjustable temples
  • Universal size