Learn More
MSC Edge Eyewear Safety Glasses, Dakura, Polycarbonate, ANSI Z87.1, 1 ea
Catalog No. 5000792368
Encompass
These Edge Eyewear Dakura safety glasses are designed for reliable eye protection. They feature a full-framed, wrap-around design with durable polycarbonate lenses, making them suitable for various industrial and laboratory environments.
  • Scratch-resistant lens coating
  • Smoke-tinted polycarbonate lenses
  • Full-framed, wrap-around design
  • Nylon frame material
  • Non-adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-007-92368 Supplier MSC Supplier No. 81945941
These Edge Eyewear Dakura safety glasses are designed for reliable eye protection. They feature a full-framed, wrap-around design with durable polycarbonate lenses, making them suitable for various industrial and laboratory environments.
  • Scratch-resistant lens coating
  • Smoke-tinted polycarbonate lenses
  • Full-framed, wrap-around design
  • Nylon frame material
  • Non-adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards