Learn More
MSC Edge Eyewear Safety Glasses, Kazbek Series, Smoke Lens, 1 ea
Catalog No. 5000768658
Encompass
These safety glasses and goggles are designed for use in bright light conditions, featuring an all-around smoke tint that blocks sunrays and glare without color distortion. They incorporate a military-grade 'Vapor Shield' anti-fog coating, ensuring clear vision even in extreme environments.
  • Interchangeable design allows easy switching between temples and a strap
  • Gloss black nylon frame
  • Soft TPR temple tips for comfort
  • Adjustable TPR nose pads
  • Allows 16% light transmission
  • Compliant with ANSI Z87.1+2010 and Military Ballistic MCEPS GL-PD 10-12 standards
Catalog No. 50-007-68658 Supplier MSC Supplier No. 55692537
These safety glasses and goggles are designed for use in bright light conditions, featuring an all-around smoke tint that blocks sunrays and glare without color distortion. They incorporate a military-grade 'Vapor Shield' anti-fog coating, ensuring clear vision even in extreme environments.
  • Interchangeable design allows easy switching between temples and a strap
  • Gloss black nylon frame
  • Soft TPR temple tips for comfort
  • Adjustable TPR nose pads
  • Allows 16% light transmission
  • Compliant with ANSI Z87.1+2010 and Military Ballistic MCEPS GL-PD 10-12 standards