Learn More
MSC Edge Eyewear Safety Glasses, Vapor Shield, Polycarbonate Lenses, 1 ea
Catalog No. 5000767382
Encompass
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses. They have a frameless, wrap-around design with a black frame and non-adjustable black temples, providing improved visibility in various conditions.
  • Meets ANSI Z87.1+ standards
  • Offers UV protection
  • Universal size
  • Nylon frame material
  • Flexible temple type
  • Anti-fog coatings improve visibility under hot and humid conditions
Catalog No. 50-007-67382 Supplier MSC Supplier No. 54308077
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses. They have a frameless, wrap-around design with a black frame and non-adjustable black temples, providing improved visibility in various conditions.
  • Meets ANSI Z87.1+ standards
  • Offers UV protection
  • Universal size
  • Nylon frame material
  • Flexible temple type
  • Anti-fog coatings improve visibility under hot and humid conditions