Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ EDGE™ Integrated Electrophoresis System
SDPSCI_ED

Catalog No. S52000 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S52000 1 to 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52000 Supplier Edvotek™ Supplier No. 500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Revolutionizes biotechnology by merging an electrophoresis chamber, power supply and blue light into a single, safe and easy-to-use apparatus.

  • Enables real-time visualization of DNA migration
  • Integrated Electrophoresis Chamber, Power Supply & Blue Light
  • Dual 100/150 Volt Modes w/ Automatic Safety Cutoff Switch
  • Includes 10 x 7 cm Gel Tray, Rubber End Caps & (2) 6/8 Tooth Combs
  • Permanent Platinum Electrodes - No Replacement Required
  • Built-in Fan & Ventilated Design Prevent Condensation
  • Compatible with SYBR™ Safe, GelGreen™ & Other Blue Light-Reactive Dyes
  • Universal Voltage for Worldwide Use
  • 3 Year Warranty

Specifications

Grade 8 to Undergraduate
Age 14 to 20
For Use With (Application) DNA & Dye Electrophoresis
Includes 10 x 7 cm Gel Tray, Rubber End Caps & (2) 6/8 Tooth Combs
Class Size 1 to 2 Lab Groups
Product Type Electrophoresis System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.