Learn More
Epredia™ Edge-Rite™ Disposable Microtome Blades
Catalog No. 1407060 Shop All Epredia Products
Change view
Click to view available options
Profile:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile
14-070-60 Low
14-070-61 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-070-60 Supplier Epredia™ Supplier No. 4280L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge-Rite™ disposable microtome blades.
  • Surgical-grade steel
  • High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
  • Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
  • Blades are 5% longer, allowing for more sections per blade
  • Blades fit any low-profile blade holder
Specifications
Product Type Disposable Microtome Blade
Profile Low
Material Surgical-grade Steel
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov