Learn More
Epredia™ Edge-Rite™ Disposable Microtome Blades
Catalog No. 1407061 Shop All Epredia Products
Change view
Click to view available options
Profile:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile
14-070-61 High
14-070-60 Low
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-070-61 Supplier Epredia™ Supplier No. 4275H
This item is not returnable. View return policy
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge-Rite™ disposable microtome blades.
  • Surgical-grade steel
  • High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
  • Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
  • Blades are 5% longer, allowing for more sections per blade
  • Blades fit any low-profile blade holder
Specifications
Product Type Disposable Microtome Blade
Profile High
Material Surgical-grade Steel
Disposable Single-use