Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Milwaukee Tool Edger Blade, 8 in, 5 PK, Carbon Steel

Catalog No. 502835788
Encompass

The Milwaukee 8 in. Edger Blade is engineered with durable carbon steel to withstand concrete impacts while edging. This blade is designed for longevity with a rust-resistant coating for water protection and features a Universal Fit Arbor.

  • Withstands concrete impacts
  • Long life; rust resistant coating
  • Universal Fit: Fits all 1 in. arbors
  • 8 in. diagonal length
  • Includes five 8 in. Edger Blades

Catalog No. 50-283-5788 Supplier MSC Supplier No. 57027914
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Milwaukee 8 in. Edger Blade is engineered with durable carbon steel to withstand concrete impacts while edging. This blade is designed for longevity with a rust-resistant coating for water protection and features a Universal Fit Arbor.

  • Withstands concrete impacts
  • Long life; rust resistant coating
  • Universal Fit: Fits all 1 in. arbors
  • 8 in. diagonal length
  • Includes five 8 in. Edger Blades

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.