Learn More
Contec™ Edgeless™ Sterile Mop Heads
BIOPHARMAGSA_VAGREENER_CHOICE
Catalog No. 191303456 Shop All Contec Inc. Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Description:
Edgeless Mop Head
Edgeless Mop Head, Ribbed Surface
Ribbed Strand Mop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Description
19-130-3456 Sterile Edgeless Mop Head, Ribbed Surface
19-042-770 Sterile Edgeless Mop Head
19-038-560 Non-sterile Ribbed Strand Mop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-3456 Supplier Contec™ Supplier No. ELMOP3IR
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Manufactured from a continuous tube of strong, durable 100% polyester knitted fabric. Edgeless™ Sterile Mop Heads are looped to form tubular mop strands that will not shred, break or deteriorate, eliminating contamination.
The Contec™ Edgeless™ Mopping System is the cleanest floor mopping system available for critical environments. The system is designed for semiconductor, pharmaceutical, biotech and medical device industries for lab clean up and process area cleaning. Mop heads are designed to meet general purpose cleaning needs in ISO Class 3 or less critical environments and are available in a wide variety of models to meet different cleaning needs. Edgeless™ or QuickConnect™ handles, frames, adaptors and bucket systems are available separately to complete the system.
  • Mop strands encapsulate liquids for maximum sorption; generate significantly lower particulate and extractable levels compared to conventional cotton string or nylon mops
  • Validated sterile per the AAMI/ISO Guidelines to a 10-6 Sterility Assurance Level (certificates of sterility available)
  • Mop heads laundered and packaged in an ISO Class 4 cleanroom
  • Resistant to chemicals, mold and mildew
  • Loops eliminate cut edges and provide improved cleanliness
  • Tail bands improved control and allow easy mopping around equipment legs and wheels
  • Available in three models:
    • Original
    • Textured--twisted strands increase sorbent capacity
    • Long textured--holds more liquid for mopping larger areas; lasts up to 10x longer than traditional mops; feature colored tail bands for area segregation to minimize cross-contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Mop Head
Sterility Sterile
Autoclavable Autoclavable
Disposable Limited-use
Description Edgeless Mop Head, Ribbed Surface
Material Polyester