Learn More
Aviva Systems Biology Corp EDIL3 Antibody - N-terminal region (ARP52090_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52090_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDIL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EDIL3. Alternate names: DEL1, MGC26287ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-633