Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Edit-R dCas9-VPR mRNA | 20 ug
Catalog No. NC1734011
Encompass
The CRISPR activation (CRISPRa) system is a variation of the canonical CRISPR-Cas9 used for creation of double-strand breaks in genomic DNA. It utilizes a nuclease-deactivated S. pyogenes Cas9 (dCas9), often called "dead Cas9", that is fused to one or more transcriptional activators. When paired with a well-designed guide RNA that targets a gene near a promoter region, the gene's native transcription start site is activated. Edit-R dCas9-VPR mRNA expresses a human codon-optimized version of the nuclease-deactivated S. pyogenes Cas9 gene, the three transcriptional activators (VP64, p65 and Rta), and two nuclear localization signals (NLS).
Catalog No. NC1734011 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. CAS12024
The CRISPR activation (CRISPRa) system is a variation of the canonical CRISPR-Cas9 used for creation of double-strand breaks in genomic DNA. It utilizes a nuclease-deactivated S. pyogenes Cas9 (dCas9), often called "dead Cas9", that is fused to one or more transcriptional activators. When paired with a well-designed guide RNA that targets a gene near a promoter region, the gene's native transcription start site is activated. Edit-R dCas9-VPR mRNA expresses a human codon-optimized version of the nuclease-deactivated S. pyogenes Cas9 gene, the three transcriptional activators (VP64, p65 and Rta), and two nuclear localization signals (NLS).