Learn More
ILEX LIFE SCIENCES LLC Immuchrom EDN (Eosinophil Derived Neurotoxin) ELISA Kit for Human Stool Samples, 96 wells
SDP
Supplier:  ILEX LIFE SCIENCES LLC IC6500
Encompass_Preferred
The ImmuChrom EDN ELISA Kit is an immunoassay that enables easy, rapid and precise quantitative determination of EDN (eosinophil derived neurotoxin) in human stool samples. Calibration Range: 0.55 ng/ml - 35 ng/ml. Detection Limit: 0.025 ng/ml. Principle: The test determines human EDN according to the sandwich-principle. EDN in sample, standard and controls binds to polyclonal antibodies, which are coated to the microtiter plate. After a washing step a peroxidase labeled polyclonal antibody is added. A second washing step is followed by the addition of the substrate which is converted to a colored product by the peroxidase. The reaction is terminated by the addition of an acidic stop solution. The optical densities are read at 450 nm in a microtiter plate reader. The EDN concentration can be calculated from the standard curve. Synonyms: eosinophil protein X, EPX, non-secretory ribonuclease, RNase UpI-2, Ribonuclease 2, RNase 2, Ribonuclease US, RNASE2, faecal EDN, fecal EDN.
Catalog No. 50-212-0004