Learn More
Aviva Systems Biology Corp EDN2 Antibody - N-terminal region (ARP52003_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52003_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDN2. It was validated on Western Blot. EDN2,endothelin 2,ET2; PPET2; EDN2,endothelin-2,endothelin-2,ET-2,preproendothelin 2,preproendothelin-2The immunogen is a synthetic peptide directed towards the n terminal region of human EDN2. Alternate names: ET2, PPET2ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-285-622