Learn More
Aviva Systems Biology Corp EDN3 Antibody - C-terminal region (ARP61173_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61173_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDN3. It was validated on Western Blot. . Alternate names: ET3, MGC15067, MGC61498, ET-3, WS4B, HSCR4, PPET3ApplicationWB. Species reactivity: Human
Catalog No. 50-287-282