Learn More
Aviva Systems Biology Corp EDNRA Antibody - C-terminal region (ARP64011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDNRA. It was validated on Western Blot. . Alternate names: ETA, ETAR, ETRA, ET-A, ETA-R, hET-ARApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Sheep
Catalog No. 50-288-141