Learn More
Medchemexpress LLC HY-101780 1mg Medchemexpress, EDO-S101 CAS:1236199-60-2 Purity:>98%
SDP
Catalog No. 501932213
Encompass_Preferred
Medchemexpress, HY-101780 1mg EDO-S101 CAS:1236199-60-2 EDO-S101 is a pan HDAC inhibitor; inhibits HDAC1, HDAC2 and HDAC3 with IC 50 values of 9, 9 and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2213 Supplier Medchemexpress LLC Supplier No. HY1017801MG
Medchemexpress, HY-101780 1mg EDO-S101 CAS:1236199-60-2 EDO-S101 is a pan HDAC inhibitor; inhibits HDAC1, HDAC2 and HDAC3 with IC 50 values of 9, 9 and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.