Learn More
Medchemexpress LLC Edonerpic maleate | 519187-97-4 | 99.8% | 407.48 | 1 ML
SDP
Catalog No. 5000387288
Encompass_Preferred
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). It is for research use only and not sold to patients.
  • Inhibits amyloid-β peptides (Aβ).
  • Preserves cortical neurons in the presence of Aβ(1-42).
  • Prevents oxidative stress-induced neuronal death at 0.1 and 1 μM.
  • Almost completely prevents GSH reduction at 0.1 and 1 μM.
  • Significantly increases neurite length at 0.1 and 1 μM in hippocampal slices.
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats infused with amyloid-beta peptide.
Catalog No. 50-003-87288 Supplier Medchemexpress LLC Supplier No. HY17631A10MM/1ML
Add to Cart
Add to Cart
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). It is for research use only and not sold to patients.
  • Inhibits amyloid-β peptides (Aβ).
  • Preserves cortical neurons in the presence of Aβ(1-42).
  • Prevents oxidative stress-induced neuronal death at 0.1 and 1 μM.
  • Almost completely prevents GSH reduction at 0.1 and 1 μM.
  • Significantly increases neurite length at 0.1 and 1 μM in hippocampal slices.
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats infused with amyloid-beta peptide.