Learn More
Medchemexpress LLC Edonerpic (maleate) | 519187-97-4 | 99.8% | 407.48 | 100 MG
SDP
Catalog No. 5000387311
Encompass_Preferred
Edonerpic maleate is a novel neurotrophic agent that can inhibit amyloid-β peptides (Aβ). It is intended for research use only and is not sold to patients. In vitro, it preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and promotes neurite outgrowth. In vivo, it ameliorates deficits in adult neurogenesis and spatial memory.
  • Novel neurotrophic agent
  • Inhibits amyloid-β peptides (Aβ)
  • In vitro efficacy: preserves cortical neurons
  • Prevents oxidative stress-induced neuronal death
  • Promotes neurite outgrowth
  • In vivo efficacy: ameliorates deficits in adult neurogenesis and spatial memory
Catalog No. 50-003-87311 Supplier Medchemexpress LLC Supplier No. HY17631A100MG
Edonerpic maleate is a novel neurotrophic agent that can inhibit amyloid-β peptides (Aβ). It is intended for research use only and is not sold to patients. In vitro, it preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and promotes neurite outgrowth. In vivo, it ameliorates deficits in adult neurogenesis and spatial memory.
  • Novel neurotrophic agent
  • Inhibits amyloid-β peptides (Aβ)
  • In vitro efficacy: preserves cortical neurons
  • Prevents oxidative stress-induced neuronal death
  • Promotes neurite outgrowth
  • In vivo efficacy: ameliorates deficits in adult neurogenesis and spatial memory