Learn More
Medchemexpress LLC Edonerpic (maleate) | 519187-97-4 | 99.8% | 407.48 | 200 MG
SDP
Catalog No. 5000387310
Encompass_Preferred
Edonerpic maleate is a novel neurotrophic agent that can inhibit amyloid-β peptides (Aβ). It preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and significantly increases neurite length. It also ameliorates deficits in adult neurogenesis and spatial memory.
  • Novel neurotrophic agent
  • Inhibits amyloid-β peptides (Aβ)
  • Preserves cortical neurons in the presence of Aβ(1-42)
  • Prevents oxidative stress-induced neuronal death at 0.1 and 1 μM
  • Almost completely prevents GSH reduction at 0.1 and 1 μM
  • Significantly increases neurite length at 0.1 and 1 μM
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats infused i.c.v. with amyloid-beta peptide
Catalog No. 50-003-87310 Supplier Medchemexpress LLC Supplier No. HY17631A200MG
Edonerpic maleate is a novel neurotrophic agent that can inhibit amyloid-β peptides (Aβ). It preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and significantly increases neurite length. It also ameliorates deficits in adult neurogenesis and spatial memory.
  • Novel neurotrophic agent
  • Inhibits amyloid-β peptides (Aβ)
  • Preserves cortical neurons in the presence of Aβ(1-42)
  • Prevents oxidative stress-induced neuronal death at 0.1 and 1 μM
  • Almost completely prevents GSH reduction at 0.1 and 1 μM
  • Significantly increases neurite length at 0.1 and 1 μM
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats infused i.c.v. with amyloid-beta peptide