Learn More
Medchemexpress LLC Edonerpic (maleate) | 519187-97-4 | 99.8% | 407.48 | 25 MG
SDP
Catalog No. 5000384074
Encompass_Preferred
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). It has been shown to preserve cortical neurons in the presence of Aβ(1-42) and prevent oxidative stress-induced neuronal death.
  • Prevents oxidative stress-induced neuronal death.
  • Almost completely prevents GSH reduction.
  • Significantly increases neurite length in hippocampal slices.
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats.
Catalog No. 50-003-84074 Supplier Medchemexpress LLC Supplier No. 25MGHY17631A
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). It has been shown to preserve cortical neurons in the presence of Aβ(1-42) and prevent oxidative stress-induced neuronal death.
  • Prevents oxidative stress-induced neuronal death.
  • Almost completely prevents GSH reduction.
  • Significantly increases neurite length in hippocampal slices.
  • Ameliorates deficits in adult neurogenesis and spatial memory in rats.