Learn More
Medchemexpress LLC Edonerpic (maleate) | 519187-97-4 | 99.8% | 407.48 | 500 MG
SDP
Catalog No. 5000387103
Encompass_Preferred
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). Supplied as a white to off-white solid, it has a high purity of 99.79%. In vitro studies show it preserves cortical neurons, prevents oxidative stress-induced neuronal death, and promotes neurite outgrowth. In vivo research indicates it ameliorates deficits in adult neurogenesis and spatial memory in amyloid-beta peptide-infused rats.
  • Neurotrophic agent
  • Amyloid-β inhibition
  • Neuronal protection: preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and inhibits GSH reduction
  • Promotes neurite outgrowth: significantly enhances neurite length in hippocampal slices
  • Cognitive and neurogenesis improvement: ameliorates deficits in adult neurogenesis and spatial memory in rats infused with amyloid-beta peptide
  • High purity: 99.79%
Catalog No. 50-003-87103 Supplier Medchemexpress LLC Supplier No. HY17631A500MG
Edonerpic maleate is a novel neurotrophic agent that inhibits amyloid-β peptides (Aβ). Supplied as a white to off-white solid, it has a high purity of 99.79%. In vitro studies show it preserves cortical neurons, prevents oxidative stress-induced neuronal death, and promotes neurite outgrowth. In vivo research indicates it ameliorates deficits in adult neurogenesis and spatial memory in amyloid-beta peptide-infused rats.
  • Neurotrophic agent
  • Amyloid-β inhibition
  • Neuronal protection: preserves cortical neurons in the presence of Aβ(1-42), prevents oxidative stress-induced neuronal death, and inhibits GSH reduction
  • Promotes neurite outgrowth: significantly enhances neurite length in hippocampal slices
  • Cognitive and neurogenesis improvement: ameliorates deficits in adult neurogenesis and spatial memory in rats infused with amyloid-beta peptide
  • High purity: 99.79%