Learn More
Cayman Chemical EdonrpIcmaleate 10mg
SDP
Catalog No. 502911889
Encompass_Preferred
A neurotrophic agent; protects against sodium nitroprusside-induced mitochondrial dysfunction and apoptosis in primary rat cortical neurons at 0.1 and 1 µM; prevents Aβ40-induced granule cell loss and improves performance in the distance movement, random reward place search, and place learning tasks in rats at 8.4 mg/kg; increases hippocampal neurogenesis and ameliorates Aβ40-induced spatial learning impairments in a rat model of Alzheimer’s disease
Catalog No. 50-291-1889 Supplier Cayman Chemical Supplier No. 3602610 MG
A neurotrophic agent; protects against sodium nitroprusside-induced mitochondrial dysfunction and apoptosis in primary rat cortical neurons at 0.1 and 1 µM; prevents Aβ40-induced granule cell loss and improves performance in the distance movement, random reward place search, and place learning tasks in rats at 8.4 mg/kg; increases hippocampal neurogenesis and ameliorates Aβ40-induced spatial learning impairments in a rat model of Alzheimer’s disease