Learn More
Medchemexpress LLC Edoxaban tosylate monohydrate | 1229194-11-9 | MFCD28400751 | 100.0% | 738.27 g·mol⁻¹ | C31H40ClN7O8S2 | 10 MM 1 ML
SDP
Catalog No. 5000245504
Encompass_Preferred
Edoxaban tosylate monohydrate is the tosylate monohydrate salt form of edoxaban, a direct factor Xa inhibitor. Supplied primarily as a 10 mM solution in 1 mL DMSO for research use, the compound is also offered as a solid in multiple milligram pack sizes and is provided with supporting documentation for safe handling and experimental use.
  • Direct factor Xa inhibitor used in anticoagulant research
  • Supplied as 10 mM solution in 1 mL DMSO for convenient stock preparation
  • Also available as solid in multiple mg pack sizes for dosing flexibility
  • High purity suitable for biochemical assays (purity reported ~99.97%)
  • Includes datasheet and SDS with molecular and safety information
Catalog No. 50-002-45504 Supplier Medchemexpress LLC Supplier No. HY10264B10MM 1ML
Edoxaban tosylate monohydrate is the tosylate monohydrate salt form of edoxaban, a direct factor Xa inhibitor. Supplied primarily as a 10 mM solution in 1 mL DMSO for research use, the compound is also offered as a solid in multiple milligram pack sizes and is provided with supporting documentation for safe handling and experimental use.
  • Direct factor Xa inhibitor used in anticoagulant research
  • Supplied as 10 mM solution in 1 mL DMSO for convenient stock preparation
  • Also available as solid in multiple mg pack sizes for dosing flexibility
  • High purity suitable for biochemical assays (purity reported ~99.97%)
  • Includes datasheet and SDS with molecular and safety information