Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Edoxaban (tosylate monohydrate) | 1229194-11-9 | MFCD28400751 | >98.0% | 738.27 | C31H40ClN7O8S2 | 100 MG
SDP

Catalog No. 5000246435
Encompass_Preferred

Edoxaban tosylate monohydrate is the tosylate monohydrate salt of edoxaban, a potent and selective direct Factor Xa (FXa) inhibitor used in anticoagulation research. Supplied as a solid reagent for laboratory and pharmacological studies; not for human or veterinary use.

  • Solid form suitable for laboratory handling.
  • Molecular formula C31H40ClN7O8S2, molecular weight 738.27 g/mol.
  • Typical storage: 4°C, sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research and reference testing involving Factor Xa inhibition.
  • Safety data and handling precautions available in supplier SDS.

Catalog No. 50-002-46435 Supplier Medchemexpress LLC Supplier No. HY10264B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Edoxaban tosylate monohydrate is the tosylate monohydrate salt of edoxaban, a potent and selective direct Factor Xa (FXa) inhibitor used in anticoagulation research. Supplied as a solid reagent for laboratory and pharmacological studies; not for human or veterinary use.

  • Solid form suitable for laboratory handling.
  • Molecular formula C31H40ClN7O8S2, molecular weight 738.27 g/mol.
  • Typical storage: 4°C, sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research and reference testing involving Factor Xa inhibition.
  • Safety data and handling precautions available in supplier SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.