Learn More
Selleck Chemical LLC Edoxaban tosylate Monohydrate
SDP
Catalog No. 5000004339
Encompass_Preferred
Edoxaban (DU-176b) is a selective factor Xa inhibitor with Ki of 0 561 nM 10 000-fold selectivity over thrombin and FIXa and is also an orally bioavailable anticoagulant drug
Catalog No. 50-000-04339 Supplier Selleck Chemical LLC Supplier No. S728050MG
Edoxaban (DU-176b) is a selective factor Xa inhibitor with Ki of 0 561 nM 10 000-fold selectivity over thrombin and FIXa and is also an orally bioavailable anticoagulant drug